Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Dehydroacetate, 98-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18606477 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-477 125 g Poly Bottle
18-606-478 500 g Poly Bottle
18-606-479 2.5 kg Poly Bottle
18-606-480 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-606-477 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S1265125GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

S1265, 64039-28-7, C8H7NaO4·H2O

Spectrum™ Chemical Sodium Dehydroacetate

Chemical Identifiers

CAS 64039-28-7
Molecular Formula C8H9NaO5
Molecular Weight (g/mol) 208.15
InChI Key AFIQQQWSAQPUQO-UHFFFAOYSA-N
IUPAC Name sodium 3-acetyl-6-methyl-2,4-dioxo-3,4-dihydro-2H-pyran-3-ide hydrate
SMILES O.[Na+].CC(=O)[C-]1C(=O)OC(C)=CC1=O

Specifications

CAS Max % 1
Packaging Poly Bottle
Quantity 125 g
Percent Purity 98.0 to 100.5%
Grade Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.