Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Dehydroepiandrosterone-3-sulfate Hydrate, TCI America™
SDP

Catalog No. S05205G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S05205G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S05205G Supplier TCI America Supplier No. S05205G

Chemical Identifiers

CAS 1099-87-2
Molecular Formula C19H29NaO6S
Molecular Weight (g/mol) 408.485
InChI Key SQMZHJFLFXVQQR-UKZXZVQVSA-M
Synonym Dehydroepiandrosterone-3-sulfate Sodium Salt, Sodium Dehydroisoandrosterone-3-sulfate
PubChem CID 23681135
IUPAC Name sodium;[(3S,8R,9S,10R,13S,14S)-10,13-dimethyl-17-oxo-1,2,3,4,7,8,9,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthren-3-yl] sulfate;hydrate
SMILES CC12CCC3C(C1CCC2=O)CC=C4C3(CCC(C4)OS(=O)(=O)[O-])C.O.[Na+]

Specifications

Melting Point 154°C
Color White
Quantity 5 g
Formula Weight 390.47
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Dehydroepiandrosterone-3-sulfate Hydrate
RTECSNumber BV8397400
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.