Learn More
Bioworld Sodium Deoxycholate, 10%, 302-95-4, 50ml
Supplier:  Bioworld 404300181
Encompass_Preferred
An anionic detergent extracted from bovine bile. High Purity Grade sodium deoxycholic acid dissolved at 10% stock solution. Limited shelf life. An ionic detergent for the solubilization of membrane-bound proteins. Keep away from light. Molecular Weight : 414.55 g/mol, Molecular Formula : C24H39O4Na,Purity : High purity grade, Solubility : Soluble in water (330 g/l) at 15 C, DMSO, methanol, and ethanol (very slightly), Appearance Form : Liquid, Appearance Color : Clear, Melting Point : 357-365 C, 50ml.
Catalog No. 50-255-883