Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium deuteroxide, for NMR, 40 wt% solution in D2O, 99+ atom % D

Catalog No. AC175470100
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Plastic Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC175470100 10 g Plastic Bottle
AC175470500 50 g Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC175470100 Supplier Thermo Scientific Chemicals Supplier No. 175470100
Add to Cart

CAS: 14014-06-3 | HNaO | 41.003 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14014-06-3 , 7789-20-0
Molecular Formula HNaO
Molecular Weight (g/mol) 41.003
MDL Number MFCD00037669
InChI Key HEMHJVSKTPXQMS-DYCDLGHISA-M
Synonym sodium deuteroxide, sodium hydroxide na od, sodium deuteroxide, 40 wt% in deuterium oxide, naod, sodium hydroxide-d, sodium deuteroxide solution, sodium deuteroxide, 1 m in deuterium oxide, sodium deuteroxide, 0.1 m in deuterium oxide, sodium deuteroxide, 30 wt% in deuterium oxide, sodium hydroxide na od 6ci,7ci,8ci,9ci
PubChem CID 23676750
SMILES [OH-].[Na+]

Specifications

Physical Form Liquid
Packaging Plastic Bottle
Quantity 10 g
Refractive Index 1.4039
Color Colorless
Deuterium (D) 99.0% min.
Density 1.4600g/mL
Linear Formula NaOD
Formula Weight 41
Grade NMR
Chemical Name or Material Sodium deuteroxide, 99+ atom % D
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 237-825-2

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.