Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Diatrizoate, MP Biomedicals™

Catalog No. MP210290850 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP210290850 50 g
MP210290810 10 g
MP210290880 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP210290850 Supplier MP Biomedicals, Inc Supplier No. 0210290850
Add to Cart
Add to Cart

Used as a density gradient reagent in blood cell separation. Sodium Diatrizoate, MP Biomedicals is also utilized as a radiological contrast agent. It causes partial redistribution of tight-junction-associated membrane proteins into a cytoplasmic compartme

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 737-31-5
Molecular Formula C11H8I3N2NaO4
Molecular Weight (g/mol) 635.898
InChI Key ZEYOIOAKZLALAP-UHFFFAOYSA-M
Synonym diatrizoate sodium, sodium diatrizoate, sodium amidotrizoate, hypaque sodium, triombrin, triombrine, hypaque, vascoray, hypaque cysto, hpaque-cysto
PubChem CID 23672589
ChEBI CHEBI:53692
IUPAC Name sodium;3,5-diacetamido-2,4,6-triiodobenzoate
SMILES CC(=O)NC1=C(C(=C(C(=C1I)C(=O)[O-])I)NC(=O)C)I.[Na+]

Specifications

Physical Form Powder
Quantity 50 g
Solubility Information Soluble in water (350mg/mL).
Formula Weight 635.9
Percent Purity >99%
Recommended Storage Room Temperature (15–30°C), protect from light.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.