Learn More
Medchemexpress LLC HY-Y0445A 100mg Medchemexpress, Sodium dichloroacetate CAS:2156-56-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYY0445A100MG
Encompass_Preferred
Medchemexpress, HY-Y0445A 100mg Sodium dichloroacetate CAS:2156-56-1 Sodium dichloroacetate is a metabolic regulator in cancer cells' mitochondria with anticancer activity. Sodium dichloroacetate inhibits PDHK, resulting in decreased lactic acid in the tumor microenvironment. Sodium dichloroacetate increases reactive oxygen species (ROS) generation and promotes cancer cell apoptosis. Sodium dichloroacetate also works as NKCC inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0763