Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodium Dichloroisocyanurate

Catalog No. 21892825G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-892-825G 25 g
21-892-8500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 21-892-825G Supplier Sigma Aldrich Supplier No. 21892825G
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Sodium dichloroisocyanurate (NaDCC) is widely used as a disinfectant for the treatment of drinking water. It is also used as a precursor to produce hypochlorous acid, a known oxidizing agent used in organic synthesis.Reactant or reagent for:N-monochlorination and dehydrochlorination of amino estersChlorination to detect ammonium via formation of colored zebra-bands in a deteting tubeSynthesis of antimicrobial polyurethane additives to control biofilmPreparation of bromochloroisocyanurate for electrophilic cobromination of alkenes and bromination of arenesStudies in regard to its antimicrobial activity on fresh produce and for emergency disinfectant of contaminated drinking water

Specifications

CAS 2893-78-9
Molecular Formula C3Cl2N3NaO3
Linear Formula C3Cl2N3NaO3
MDL Number MFCD00036333
Quantity 25 g
Synonym NaDCC; 3,5-Dichloro-2-hydroxy-4,6-s-triazinedione sodium salt; Dichloro-s-triazinetrione sodium salt
Molecular Weight (g/mol) 219.95 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.