Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Dichromate Dihydrate, Reagent Grade, Innovating Science™
SCI_ED

Catalog No. S000244 Shop All Aldon Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S000244 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING: This product can expose you to chemicals including Chromium, hexavalent compounds, which are known to the State of California to cause cancer and reproductive harm. For more information go to www.P65Warnings.ca.gov.
Catalog No. S000244 Supplier Aldon Corporation Supplier No. IS28105
Add to Cart
Add to Cart
WARNING: This product can expose you to chemicals including Chromium, hexavalent compounds, which are known to the State of California to cause cancer and reproductive harm. For more information go to www.P65Warnings.ca.gov.

Chemical Identifiers

Molecular Weight (g/mol) 298.05 g/mol

Specifications

CAS 7789-12-0
Physical Form Crystals
Quantity 500 g
Percent Purity 100%
Grade Reagent
Packaging HDPE
Chemical Name or Material Sodium dichromate, dihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.