Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodium Dicyanamide

Catalog No. 6903091KG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
69-030-91KG 20 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 69-030-91KG Supplier Sigma Aldrich Supplier No. 6903091KG
May include imposed supplier surcharges.
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reactant for preparation of:Nickel diacetylpyridine dioximate pentanuclear, octanuclear and decanuclear cluster complexesDecanuclear manganese(II/III) oxo azide diolate clusters as supertetrahedral building-blocks to design extended magnetic networksRuthenium bipyridine complexes entrapped in iron and manganese dicyanamide two-dimensional anionic sheet-like coordination polymeric networks exhibiting dual emission photoluminescenceLead dicyanamide-bridged nitronyl nitroxide radical polymeric complexAntiferromagnetically coupled hexanuclear copper(II) Schiff base complexes with dicyanamido bridgesCuII2L based polymeric ladders using dicyanamide bridgesLuminous hexacoordinated coordination polymers of cadmium(II), containing dicyanamide bridges

Specifications

CAS 7727-43-7
Molecular Formula BaO4S
Linear Formula BaSO4
MDL Number MFCD00003455
Quantity 20 g
Molecular Weight (g/mol) 233.39 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.