Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodium Dicyanamide

Catalog No. 17832225G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-832-225G 25 g
17-832-2100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 17-832-225G Supplier Sigma Aldrich Supplier No. 17832225G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reactant for preparation of:Nickel diacetylpyridine dioximate pentanuclear, octanuclear and decanuclear cluster complexesDecanuclear manganese(II/III) oxo azide diolate clusters as supertetrahedral building-blocks to design extended magnetic networksRuthenium bipyridine complexes entrapped in iron and manganese dicyanamide two-dimensional anionic sheet-like coordination polymeric networks exhibiting dual emission photoluminescenceLead dicyanamide-bridged nitronyl nitroxide radical polymeric complexAntiferromagnetically coupled hexanuclear copper(II) Schiff base complexes with dicyanamido bridgesCuII2L based polymeric ladders using dicyanamide bridgesLuminous hexacoordinated coordination polymers of cadmium(II), containing dicyanamide bridges

Specifications

CAS 1934-75-4
Molecular Formula C2N3Na
Linear Formula NaN(CN)2
MDL Number MFCD00003535
Quantity 25 g
Molecular Weight (g/mol) 89.03 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.