Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodium Diformylamide

Catalog No. 7149450G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-494-50G 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 71-494-50G Supplier Sigma Aldrich Supplier No. 7149450G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reactant for preparation of:Functionalized condensed benzazoles+a-ketoheterocycles as inhibitors of Leishmania mexicana cysteine protease CPBINOS inhibitor PHA-399733 via Bucherer-Bergs hydantoin synthesisAntibiotic resistance (C +a-lactamase AmpC ) inhibitor analogues, with improved cell permeability propertiesBINOL- and H8-BINOL-based aryl phosphites as ligands in palladium-catalyzed asymmetric allylation and amination reactionsPrecursors of +aV+a3 integrin antagonistsModified Gabriel reagent for the synthesis of amines; mild hydrolysis to release the amine

Specifications

CAS 384842-25-5
Melting Point 130°C to 133°C
Molecular Formula C23H31P
Linear Formula C23H31P
MDL Number MFCD16621412
Quantity 500 mg
Synonym Diformylimide sodium salt
Molecular Weight (g/mol) 338.47 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.