Learn More
Strem, An Ascensus Company CAS# 10049-21-5. 250g. Sodium dihydrogen phosphate monohydrate (99.9%-Na). MFCD00149208
SDP
Catalog No. 5090113894
Encompass
CAS# 10049-21-5. 250g. Sodium dihydrogen phosphate monohydrate (99.9%-Na). MFCD00149208. Molecular Weight: 120.05 (137.99). Molecular Formula: NaH2PO4∙H2O. Color/form: white pwdr. Strem# 93-1174. http://www.strem.com/catalog/v/93-1174/
Catalog No. 50-901-13894 Supplier Strem, An Ascensus Company Supplier No. 931174250G
CAS# 10049-21-5. 250g. Sodium dihydrogen phosphate monohydrate (99.9%-Na). MFCD00149208. Molecular Weight: 120.05 (137.99). Molecular Formula: NaH2PO4∙H2O. Color/form: white pwdr. Strem# 93-1174. http://www.strem.com/catalog/v/93-1174/