Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Dihydrogen Phosphate Dihydrate, For Analysis, EMSURE™, MilliporeSigma™

Catalog No. M1063420250 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 g
Packaging:
Plastic Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1063420250 250 g Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1063420250 Supplier MilliporeSigma Supplier No. 1.06342.0250
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity reagent compliant with European Pharmacopoeia standards, suitable for classical analytical applications.

Recommended Storage

+5°C to +30°C.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13472-35-0
Molecular Formula H6NaO6P
Molecular Weight (g/mol) 156.006
InChI Key VBJGJHBYWREJQD-UHFFFAOYSA-M
Synonym sodium dihydrogen phosphate dihydrate, sodium phosphate monobasic dihydrate, monosodium phosphate dihydrate, sodium phosphate, monobasic, dihydrate, phosphoric acid, monosodium salt, dihydrate, sodium dihydrogenphosphate dihydrate, sodium phosphate nah2po4 dihydrate, monosodium dihydrogen phosphate dihydrate, sodium dihydrate dihydrogen phosphate, acmc-20ajoh
PubChem CID 23673460
IUPAC Name sodium;dihydrogen phosphate;dihydrate
SMILES O.O.OP(=O)(O)[O-].[Na+]

Specifications

Melting Point 60°C
Color Colorless
pH 4.5 (12g/L, H2O, 25°C)
Physical Form Crystals
Quantity 250 g
Solubility Information 850g/L
Formula Weight 156.02
Percent Purity 99.0 to 100.5%
Grade Reagent
Packaging Plastic Bottle
Density 1.915 g/3 (20 °C)
Chemical Name or Material Sodium Dihydrogen Phosphate Dihydrate
Show More Show Less
EINECSNumber 231-449-2
Recommended Storage +5°C to +30°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.