Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium dihydrogen phosphate monohydrate EMPROVE™ ESSENTIAL BP, USP, MilliporeSigma™

Catalog No. M1063499013 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
12 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1063499013 12 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1063499013 Supplier MilliporeSigma Supplier No. 1063499013

Chemical Identifiers

CAS 10049-21-5
Molecular Formula H2NaO4P*H2O
Molecular Weight (g/mol) 137.99

Specifications

pH 4.1 to 4.5 (50 g/L, H2O, 25°C)
Quantity 12 kg
Assay Percent Range 98.0 to 100.5%
Solubility Information 850 g/L
Packaging Plastic Bucket
Density 2.04 g/cm3 (at 20°C); Bulk Density: 880 kg/m3
Recommended Storage Store at 15 to 25°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.