Learn More
Medchemexpress LLC Sodium diphenylamine 1g
SDP
Catalog No. 5000205134
Encompass_Preferred
Sodium diphenylamine-4-sulfonate is a chemical indicator commonly used in redox titrations Its reduced state is colorless and its oxidized state is reddish-purple Sodium diphenylamine-4-sulfonate is also a quencher and reducing agent Sodium diphenylamine-4-sulfonate inhibits the electrochemiluminescence of the ruthenium tris(bipyridine) system through energy transfer Sodium diphenylamine-4-sulfonate participates in the synthesis of nanocrystals[1][2][3][4]
Catalog No. 50-002-05134 Supplier Medchemexpress LLC Supplier No. HYW1107911G
Sodium diphenylamine-4-sulfonate is a chemical indicator commonly used in redox titrations Its reduced state is colorless and its oxidized state is reddish-purple Sodium diphenylamine-4-sulfonate is also a quencher and reducing agent Sodium diphenylamine-4-sulfonate inhibits the electrochemiluminescence of the ruthenium tris(bipyridine) system through energy transfer Sodium diphenylamine-4-sulfonate participates in the synthesis of nanocrystals[1][2][3][4]