Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium disulfite (sodium metabisulfite), 100.0%, EMPROVE™ ESSENTIAL Ph Eur, BP, JP, NF, E 223, MilliporeSigma™

Catalog No. M1063575000 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1063575000 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. M1063575000 Supplier MilliporeSigma Supplier No. 1063575000
This item is not returnable. View return policy

Chemical Identifiers

CAS 7681-57-4
Molecular Formula Na2O5S2
Molecular Weight (g/mol) 190.11
Synonym Sodium metabisulfite, Sodium pyrosulfite, Disulfite, Pyrosulfite

Specifications

Melting Point 150°C (decomposition)
pH 3.5 to 5 (50 g/L, H2O, 20°C)
Quantity 5 kg
Assay Percent Range Assay (iodometric): 97.0 to 100.0%; Assay (iodometric, SO2): 65.4 to 67.4%
Solubility Information 650 g/L
Packaging Plastic Bottle
Density 2.36 g/cm3 (at 20°C); Bulk Density: 1000 to 1200 kg/m3
Recommended Storage Store at 5 to 30°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.