Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodium Dl-Lactate

Catalog No. 7172025G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-720-25G 25 g
71-720-5G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 71-720-25G Supplier Sigma Aldrich Supplier No. 7172025G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Sodium DL-lactate (DL-lactate) has been employed as a carbon source in the minimal salt medium (MSM) for the culture of Pseudomonas putida KT2440. It may be employed as an energy source in the basal complex medium for the growth of Selenomonas ruminantium strains.Contains approximately equal amounts of D and L isomers.ReagentPlus is a registered trademark of Sigma-Aldrich Co. LLC

Specifications

CAS 72-17-3
Molecular Formula C3H5NaO3
Linear Formula CH3CH(OH)COONa
MDL Number MFCD00065400
Quantity 25 g
Synonym (+/-)-2-Hydroxypropionic acid sodium salt; DL-Lactic acid sodium salt
Molecular Weight (g/mol) 112.06 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.