Learn More
Medchemexpress LLC Sodium dodecanoate 500mg
SDP
Catalog No. 5000205445
Encompass_Preferred
Sodium LaurateIt is an organic compound belonging to fatty acids It is the sodium salt of lauric acid a medium-chain saturated fatty acid found in various animal and vegetable fats Sodium LaurateCommonly used as surfactants emulsifiers and cleaning agents in various industries such as cosmetics personal care and detergent industries It s found in a variety of products including soaps shampoos toothpaste and laundry detergent Furthermore it also has applications in the food industry where it is used as a food additive to improve texture and stability
Catalog No. 50-002-05445 Supplier Medchemexpress LLC Supplier No. HYW099601500MG
Sodium LaurateIt is an organic compound belonging to fatty acids It is the sodium salt of lauric acid a medium-chain saturated fatty acid found in various animal and vegetable fats Sodium LaurateCommonly used as surfactants emulsifiers and cleaning agents in various industries such as cosmetics personal care and detergent industries It s found in a variety of products including soaps shampoos toothpaste and laundry detergent Furthermore it also has applications in the food industry where it is used as a food additive to improve texture and stability