Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium dodecatungstophosphate hydrate

Catalog No. AAA1157514
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1157514 25 g
AAA1157522 100 g
AAA1157536 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1157514 Supplier Thermo Scientific Chemicals Supplier No. A1157514
Add to Cart

CAS: 51312-42-6 | Na3O40PW12 | 2946.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51312-42-6
Molecular Formula Na3O40PW12
Molecular Weight (g/mol) 2946.12
MDL Number MFCD00036311
IUPAC Name Sodium dodecamolybdophosphate hydrate
SMILES *

Specifications

Melting Point 350°C
Quantity 25 g
Formula Weight 2946.12 (Anhydrous)
Chemical Name or Material Sodium dodecatungstophosphate hydrate
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 257-132-9
RTECSNumber TH5775000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.