Learn More
Medchemexpress LLC Sodium dodecyl sulfate | 151-21-3 | MFCD00036175 | 98.0% | 288.38 g/mol | C12H25NaO4S | 5g
SDP
Catalog No. 5000265894
Encompass_Preferred
Sodium dodecyl sulfate for electrophoresis is a high purity sodium dodecyl sulfate (SDS) with a minimum purity of 98 5% It is an anionic detergent commonly used in electrophoretic procedures to denature and impart a negative charge to proteins allowing separation based on size and charge Sodium dodecyl sulfate for electrophoresis has unique chemical properties that make it an effective ingredient in many laboratory applications helping to improve the resolution and sensitivity of protein analysis
Catalog No. 50-002-65894 Supplier Medchemexpress LLC Supplier No. HYY0316B5G
Sodium dodecyl sulfate for electrophoresis is a high purity sodium dodecyl sulfate (SDS) with a minimum purity of 98 5% It is an anionic detergent commonly used in electrophoretic procedures to denature and impart a negative charge to proteins allowing separation based on size and charge Sodium dodecyl sulfate for electrophoresis has unique chemical properties that make it an effective ingredient in many laboratory applications helping to improve the resolution and sensitivity of protein analysis