Learn More
Medchemexpress LLC Sodium dodecyl sulfate | 151-21-3 | MFCD00036175 | 99.9% | 288.38 g/mol | C12H25NaO4S | 25g
SDP
Catalog No. 5000269992
Encompass_Preferred
Sodium dodecyl sulfate is an anionic surfactant commonly used as a detergent emulsifier and protein denaturant in various industrial processes especially in the production of personal care products cleaners and laboratory reagents Sodium dodecyl sulfate has unique chemical properties that make it an effective ingredient in many applications helping to reduce surface tension and enhance cleaning power
Catalog No. 50-002-69992 Supplier Medchemexpress LLC Supplier No. HYY031625G
Sodium dodecyl sulfate is an anionic surfactant commonly used as a detergent emulsifier and protein denaturant in various industrial processes especially in the production of personal care products cleaners and laboratory reagents Sodium dodecyl sulfate has unique chemical properties that make it an effective ingredient in many applications helping to reduce surface tension and enhance cleaning power