Learn More
Bio Rad Laboratories SODIUM DODECYL SULFATE 1KG
Catalog No. NC9289190
Encompass
This highly purified SDS detergent optimizes activity recovery in enzyme purification applications. Supplied as 1 kg powder. (161-0302)
Catalog No. NC9289190 Supplier Bio Rad Laboratories Supplier No. 1610302
This highly purified SDS detergent optimizes activity recovery in enzyme purification applications. Supplied as 1 kg powder. (161-0302)