Learn More
Research Products International Corp SDS, Micro-Pellets [Sodium dodecylsulfate][Lauryl sulfate], 5 Kilograms
SDP
Catalog No. 50488741
Encompass_Preferred
New micro-pellets allow convenient weighing and handling without the mess and odor of? powders. High purity SDS is additive free. Easy to dissolve in water for denaturing proteins prior to gel elecrophoresis.
Catalog No. 50-488-741 Supplier Research Products International Corp Supplier No. L220405000.0
New micro-pellets allow convenient weighing and handling without the mess and odor of? powders. High purity SDS is additive free. Easy to dissolve in water for denaturing proteins prior to gel elecrophoresis.