Learn More
Sigma Aldrich Fine Chemicals Biosciences Sodium dodecyl sulfate, 151-21-3, MFCD00036175, 500 G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences L3771500G
Encompass
Linear Formula: CH3(CH2)11OSO3Na, Molecular Weight: 288.38, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), Synonym: Dodecyl sodium sulfate, Dodecyl sulfate sodium salt, Lauryl sulfate sodium salt, SDS, Sodium lauryl sulfate.
Catalog No. NC0540479