Learn More
Sigma Aldrich Fine Chemicals Biosciences Sodium dodecyl sulfate | 151-21-3 | MFCD00036175 | 250g
Catalog No. NC0618616
Encompass
Sodium dodecyl sulfate | Purity: ≥98.5% (GC) | MW:288.38 | 151-21-3 | MFCD00036175 | 250g
Catalog No. NC0618616 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. L4509250G
Sodium dodecyl sulfate | Purity: ≥98.5% (GC) | MW:288.38 | 151-21-3 | MFCD00036175 | 250g