Learn More
Sigma Aldrich Fine Chemicals Biosciences Sodium dodecyl sulfate, 151-21-3, MFCD00036175, 100 G
Catalog No. NC0559180
Encompass
Linear Formula: CH3(CH2)11OSO3Na, Molecular Weight: 288.38, ACS reagent, ≥99.0%, Synonym: Dodecyl sodium sulfate, Dodecyl sulfate sodium salt, Lauryl sulfate sodium salt, SDS, Sodium lauryl sulfate. Sodium dodecyl sulfate was used as a test compound for studying that whether Bacillus amyloliquefaciens KPS46 can induce priming, signaling, protein production as well as systemic protection in soybean (Glycine max L.) against several diseases.
Catalog No. NC0559180 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 436143100G
Linear Formula: CH3(CH2)11OSO3Na, Molecular Weight: 288.38, ACS reagent, ≥99.0%, Synonym: Dodecyl sodium sulfate, Dodecyl sulfate sodium salt, Lauryl sulfate sodium salt, SDS, Sodium lauryl sulfate. Sodium dodecyl sulfate was used as a test compound for studying that whether Bacillus amyloliquefaciens KPS46 can induce priming, signaling, protein production as well as systemic protection in soybean (Glycine max L.) against several diseases.