Learn More
Sigma Aldrich Fine Chemicals Biosciences Sodium dodecyl sulfate anh500G
Catalog No. 502906453
Encompass
SDS (sodium dodecyl sulfate) is a wetting agent and an anionic detergent which functions efficiently in both alkaline and acidic SOLUTIONons. It is generally used for the solubilization of lipids and proteins. It also functions as a potent protein denaturant. SDS volume-wise is one of the most commonly used synthetic surfactants.
Catalog No. 50-290-6453 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. RDD021500G
SDS (sodium dodecyl sulfate) is a wetting agent and an anionic detergent which functions efficiently in both alkaline and acidic SOLUTIONons. It is generally used for the solubilization of lipids and proteins. It also functions as a potent protein denaturant. SDS volume-wise is one of the most commonly used synthetic surfactants.