Learn More
Sigma Aldrich Fine Chemicals Biosciences Sodium dodecyl sulfate Bio10KG
Catalog No. 502907631
Encompass
Sodium dodecyl sulfate (SDS) is an anionic detergent suitable to denature proteins. It has a long-chain aliphatic group with negatively charged sulfate attached. This makes SDS an amphipathic detergent.
Catalog No. 50-290-7631 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. L377110KG
Sodium dodecyl sulfate (SDS) is an anionic detergent suitable to denature proteins. It has a long-chain aliphatic group with negatively charged sulfate attached. This makes SDS an amphipathic detergent.