Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Dodecyl Sulfate 97.0+%, TCI America™
SDP

Catalog No. S0588500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0588500G 500 g
S058825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S0588500G Supplier TCI America Supplier No. S0588500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 151-21-3
Molecular Formula C12H25NaO4S
Molecular Weight (g/mol) 288.38
MDL Number MFCD00036175
InChI Key DBMJMQXJHONAFJ-UHFFFAOYSA-M
Synonym Dodecylsulfuric Acid Sodium Salt, Laurylsulfuric Acid Sodium Salt, Sodium Lauryl Sulfate, SDS
PubChem CID 3423265
ChEBI CHEBI:8984
IUPAC Name sodium dodecyl sulfate
SMILES [Na+].CCCCCCCCCCCCOS([O-])(=O)=O

Specifications

Color White-Yellow
Physical Form Crystal-Powder at 20°C
Melting Point 207°C
Quantity 500 g
Formula Weight 288.38
Percent Purity ≥97.0% (GC,T)
Chemical Name or Material Sodium Dodecyl Sulfate [for Electrophoresis]
EINECSNumber (2)-1679
RTECSNumber WT1050000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.