Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Sodium Dodecyl Sulfate, OmniPur™, Calbiochem™,

Catalog No. 79605KG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 g
5 kg
Packaging:
Plastic Bottle
Plastic Drum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
79-605-KG 5 kg Plastic Drum
79-105-00GM 500 g Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 79-605-KG Supplier MilliporeSigma™ Supplier No. 79605KG
Add to Cart
Add to Cart

Recommended Storage

Ok to freeze

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 151-21-3
Molecular Formula C12H25NaO4S
Molecular Weight (g/mol) 288.38
MDL Number MFCD00036175
InChI Key DBMJMQXJHONAFJ-UHFFFAOYSA-M
Synonym SDS, Sodium Lauryl Sulfate, n-Dodecyl Sulfate, Sodium
PubChem CID 3423265
ChEBI CHEBI:8984
IUPAC Name sodium dodecyl sulfate
SMILES [Na+].CCCCCCCCCCCCOS([O-])(=O)=O

Specifications

Color White
Physical Form Fine powder
Quantity 5 kg
Packaging Plastic Drum
Percent Purity ≥99.0% (dried basis)
Grade Molecular Biology
Chemical Name or Material Sodium Dodecyl Sulfate
storageNote1 Ok to freeze
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.