Learn More
Research Products International Corp SDS [Sodium dodecylsulfate] [Lauryl sulfate], Powder, 1 Kilogram
SDP
Catalog No. 50550412
Encompass_Preferred
Ionic detergent for denaturing proteins prior to gel electrophoresis.
Catalog No. 50-550-412 Supplier Research Products International Corp Supplier No. L220101000.0
Ionic detergent for denaturing proteins prior to gel electrophoresis.