Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium Dodecyl Sulfate (SDS), ≥95%
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Cardboard Carton
Plastic bottle
Specifications
Specifications
Physical Form | Crystalline Powder |
Melting Point | ∼206°C |
Quantity | 1 kg |
Packaging | Plastic bottle |
Linear Formula | CH3(CH2)11OSO3Na |
UN Number | UN2926 |
Beilstein | 3599286 |
IUPAC Name | sodium;dodecyl sulfate |
Formula Weight | 288.38 |
Percent Purity | ≥95% |
Show More |
Safety and Handling
Hazard Category | H302-H315-H318-H500 |
Precautionary Statement | P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P310-P330-P332+P313-P362-P501c |
EINECSNumber | 205-788-1 |
RTECSNumber | WT1050000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction