Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Dodecyl Sulfate (Sodium Lauryl Sulfate), Reagents

Catalog No. 01337272
Change view
Click to view available options
Quantity:
100 mL
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-272 100 mL
01-337-271 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-337-272 Supplier Reagents Holdings Llc Supplier No. C2197800100A6
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium Dodecyl Sulfate (Sodium Lauryl Sulfate), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 151-21-3
Molecular Formula C12H25NaO4S
Molecular Weight (g/mol) 288.38
MDL Number MFCD00036175
InChI Key DBMJMQXJHONAFJ-UHFFFAOYSA-M
Synonym Sodium lauryl sulfate
IUPAC Name sodium dodecyl sulfate
SMILES [Na+].CCCCCCCCCCCCOS([O-])(=O)=O

Specifications

Color White
Physical Form Solid
Melting Point 204°C to 207°C
Quantity 100 mL
Packaging Poly Bottle
Specific Gravity 1.1
Chemical Name or Material Sodium Dodecyl Sulfate (Sodium Lauryl Sulfate)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.