Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Dodecyl Sulfate, Ultrapure, 500 g, Innovating Science™
GREENER_CHOICESCI_ED

Catalog No. S000980 Shop All Aldon Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S000980 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S000980 Supplier Aldon Corporation Supplier No. IS29044
Add to Cart
Add to Cart
  • Appearance: White Powder
  • Purity: ≥ 98.0%
  • Active Matter: ≥ 99%
  • Identification (FTIR): Conforms to Structure
  • Lead: ≤ 5 ppm
  • Phosphate: ≤ 1 ppm
  • Absorbance at 260 nm (3% Aqueous): ≤ 0.4
  • Absorbance at 280 nm (3% Aqueous): ≤ 0.1
  • DNase: None Detected
  • RNase: None Detected

Chemical Identifiers

CAS 151-21-3
Molecular Weight (g/mol) 288.38

Specifications

pH 9.1
Quantity 500 g
Packaging HDPE
Percent Purity 100%
Grade Ultra Pure
Chemical Name or Material Sodium doecyl sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.