Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Dodecyl Sulfate (SDS), White Powder, Electrophoresis, Fisher BioReagents™

Catalog No. p-28617 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
500 g
5 kg
Packaging:
Glass Bottle
Poly Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP166-100 100 g Glass Bottle
BP166-500 500 g Poly Bottle
BP166-5 5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. BP166-100
Only null left
Add to Cart

For protein extraction procedures and SDS polyacrylamide gels | CAS: 151-21-3 | C12H25NaO4S | 288.38 g/mol

For protein extraction procedures and SDS polyacrylamide gels

  • DNase and RNase-free detergent for protein extraction procedures and polyacrylamide gel electrophoresis
  • SDS is a commonly used detergent in protein purification and polyacrylamide gel electrophoresis (PAGE)
  • SDS denatures proteins by disrupting the non-covalent bonds of the protein
  • Functionally tested for use in electrophoresis
  • Intended for research use only; not for human or animal diagnostic purposes
TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C12H25NaO4S
Molecular Weight (g/mol) 288.38
MDL Number MFCD00036175
InChI Key DBMJMQXJHONAFJ-UHFFFAOYSA-M
Synonym Sodium Lauryl Sulfate, SDS
PubChem CID 3423265
ChEBI CHEBI:8984
IUPAC Name sodium dodecyl sulfate
SMILES [Na+].CCCCCCCCCCCCOS([O-])(=O)=O

Specifications

Absorbance 0.1 max. (0.1% solution) at 280nm, 0.4 max. (0.1% solution) at 230nm
Color White
CAS 151-21-3, 7647-14-5, 7757-82-6
Physical Form Solid
Melting Point 206°C
pH 8.5 to 10
Quantity 100 g
Identification Pass Test
Packaging Glass Bottle
Assay Percent Range (as Fatty Alcohol Sulfate) >=99%
Linear Formula CH3(CH2)10CH2OSO3Na
Merck Index 15, 8768
Formula Weight 288.38
Percent Purity ≥99% (≥98% C12-OH)
Grade DNase- and RNase-Free
Assay (as Fatty Alcohol Sulfate) >=99%
Chemical Name or Material Sodium Dodecyl Sulfate
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Flammable solid. Causes eye, skin, and respiratory tract irritation. May cause allergic respiratory reaction. Avoid contact with skin and eyes. Do not breathe dust. Use explosion-proof equipment. Use only non-sparking tools. Wash off immediately with soap and plenty of water removing all contaminated clothes and shoes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Remove from exposure, lie down. Remove from exposure, lie down. Move to fresh air.

NFPA

Health:2
Flammability:3
Instability:0
Recommended Storage RT
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.