Learn More
FUJIFILM Biosciences Sodium Dodecylbenzene Sulfonat
Catalog No. 5000450385
Encompass_Preferred
This product conforms to the Japanese Pharmacopoeia General Test Methods for Sodium Dodecylbenzenesulfonate as a reagent/solution It can be used to evaluate the performance of total organic carbon analyzers TOC analyzers when measuring organic carbon content in the Organic Carbon Test Method
Catalog No. 50-004-50385 Supplier FUJIFILM Biosciences Supplier No. 19216421
This product conforms to the Japanese Pharmacopoeia General Test Methods for Sodium Dodecylbenzenesulfonate as a reagent/solution It can be used to evaluate the performance of total organic carbon analyzers TOC analyzers when measuring organic carbon content in the Organic Carbon Test Method