Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodium dodecylbenzenesulfonate

Catalog No. 289957500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
500 g
1 kg
10 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
28-995-7500G 500 g
28-995-725G 25 g
28-995-71KG 1 kg
28-995-710KG 10 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 28-995-7500G Supplier Sigma Aldrich Supplier No. 289957500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Sodium dodecylbenzenesulfonate is an ionic surfactant.Sodium dodecylbenzenesulfonate was used to fabricate amperometic biosensors for glucose.

Specifications

CAS 25155-30-0
Molecular Formula C18H29NaO3S
Linear Formula CH3(CH2)11C6H4SO3Na
MDL Number MFCD00011508
Quantity 500 g
Synonym SDBS; Dodecylbenzenesulfonic acid sodium salt
Molecular Weight (g/mol) 348.48
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.