Learn More
Medchemexpress LLC Sodium dodecylbenzenesulfonate | 25155-30-0 | 90.2% | 1 KG
SDP
Supplier:  Medchemexpress LLC HYB19371KG
Encompass_Preferred
Sodium dodecylbenzenesulfonate is a nonionic surfactant primarily used for cleaning and treating industrial equipment. It offers good cleaning power and environmental compatibility, effectively removing oil, dirt, and other contaminants. This compound also finds applications in the textile, paper, and leather industries for fixing and stabilizing dyes and colors. Recognized for its significant role in industrial processes and laboratory research, it serves as a biochemical reagent for life science-related research.
  • Nonionic surfactant
  • Good cleaning power
  • Environmental compatibility
  • Effectively removes oil, dirt, and contaminants
  • Used in textile, paper, and leather industries for fixing and stabilizing dyes
  • Biochemical reagent for life science research
Catalog No. 50-004-11103