Learn More
Medchemexpress LLC Sodium edetate | 64-02-8 | MFCD00012460 | 98.0% | 380.17 g/mol | C10H12N2Na4O8 | 100g
SDP
Catalog No. 5000267968
Encompass_Preferred
Ethylenediaminetetraacetic acid (EDTA) tetrasodium is a kind of metal chelating agent (binds to bivalent and trivalent metal cations including calcium) Ethylenediaminetetraacetic acid tetrasodium has antibacterial anti-inflammatory antioxidant anti-hypercalcemia and anticoagulant activities Ethylenediaminetetraacetic acid tetrasodium decreases the metal ion-catalyzed oxidative damage to proteins and allows maintenance of reducing environment during protein purification Ethylenediaminetetraacetic acid tetrasodium can alleviate the liver fibrosis Ethylenediaminetetraacetic acid tetrasodium can be used for coronary artery disease and neural system disease research[1][2][3][4][5][6][7]
Catalog No. 50-002-67968 Supplier Medchemexpress LLC Supplier No. HYY0682B100G
Ethylenediaminetetraacetic acid (EDTA) tetrasodium is a kind of metal chelating agent (binds to bivalent and trivalent metal cations including calcium) Ethylenediaminetetraacetic acid tetrasodium has antibacterial anti-inflammatory antioxidant anti-hypercalcemia and anticoagulant activities Ethylenediaminetetraacetic acid tetrasodium decreases the metal ion-catalyzed oxidative damage to proteins and allows maintenance of reducing environment during protein purification Ethylenediaminetetraacetic acid tetrasodium can alleviate the liver fibrosis Ethylenediaminetetraacetic acid tetrasodium can be used for coronary artery disease and neural system disease research[1][2][3][4][5][6][7]