Learn More
Medchemexpress LLC HY-N7079A 500mg Medchemexpress, Sodium erythorbate CAS:6381-77-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN7079A500MG
Encompass_Preferred
Medchemexpress, HY-N7079A 500mg Sodium erythorbate CAS:6381-77-7 Sodium erythorbate (D-Isoascorbic acid sodium), produced from sugars derived from different sources, such as beets, sugar cane, and corn, is a food additive used predominantly in meats, poultry, and soft drinks. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2983