Learn More
Medchemexpress LLC Sodium erythorbate | 6381-77-7 | 10 MM 1 ML
SDP
Catalog No. 5000418041
Encompass_Preferred
Sodium erythorbate is an orally active sodium salt form of isoascorbic acid. It reacts with free radicals to reduce oxidative damage and has weak antiscorbic activity. It can also prevent oxidative discoloration in meat products.
  • Improves radiation resistance of *Listeria monocytogenes*
  • Reacts with free radicals and reduces oxidative damage
  • Has weak antiscorbic activity
  • Prevents oxidative discoloration in meat products
Catalog No. 50-004-18041 Supplier Medchemexpress LLC Supplier No. HYN7079A10MM/1ML
Sodium erythorbate is an orally active sodium salt form of isoascorbic acid. It reacts with free radicals to reduce oxidative damage and has weak antiscorbic activity. It can also prevent oxidative discoloration in meat products.
  • Improves radiation resistance of *Listeria monocytogenes*
  • Reacts with free radicals and reduces oxidative damage
  • Has weak antiscorbic activity
  • Prevents oxidative discoloration in meat products