Learn More
Description
- Sodium ethanethiolate is a reagent generally used to synthesize various thioethers
- It can be employed as a ligand in the preparation of organometallic complexes
- It is also used in the synthesis of thiolated amino-alcohols.alkanethiolates are efficient reagents for sn2 dealkylation of esters and aryl ethers
Specifications
Specifications
| CAS | 811-51-8 |
| Molecular Formula | C2H5NaS |
| Linear Formula | CH3CH2SNa |
| MDL Number | MFCD00066318 |
| Quantity | 50 g |
| Synonym | Ethanethiol sodium salt; Ethyl mercaptan sodium salt |
| Molecular Weight (g/mol) | 84.12 |
| Percent Purity | ∼90% (RT) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.