Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium ethoxide, 21% w/w in ethanol

Catalog No. AAL13083AE
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL13083AE 100 mL
AAL13083AP 500 mL
AAL130830F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL13083AE Supplier Thermo Scientific Chemicals Supplier No. L13083AE
Only null left
Add to Cart

CAS: 141-52-6 | C2H5NaO | 68.051 g/mol

Sodium ethoxide, 21% w/w in ethanol is used as a strong base in organic synthesis. It finds application in various chemical reactions such as condensation, esterification, alkoxylation and etherifcation. It is actively involved in Claisen condensation, Stobbe reaction and Wolf-kishner reduction. It is an important starting material for the synthesis of ethyl ester and diethyl ester of malonic acid. In Williamson ether synthesis, it reacts with ethyl bromide to form diethyl ether.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium ethoxide, 21% w/w in ethanol is used as a strong base in organic synthesis. It finds application in various chemical reactions such as condensation, esterification, alkoxylation and etherifcation. It is actively involved in Claisen condensation, Stobbe reaction and Wolf-kishner reduction. It is an important starting material for the synthesis of ethyl ester and diethyl ester of malonic acid. In Williamson ether synthesis, it reacts with ethyl bromide to form diethyl ether.

Solubility
Miscible with alcohol.

Notes
Moisture sensitive. It hydrolyzes rapidly into sodium hydroxide in the presence of moist air. Incompatible with water, strong bases, acids, oxidizing agents, alkali metals, strong oxidizing agents, ammonia, acid chlorides, acid anhydrides, reducing agents, peroxides, acids and chlorinated solvents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-52-6
Molecular Formula C2H5NaO
Molecular Weight (g/mol) 68.051
MDL Number MFCD00012417
InChI Key QDRKDTQENPPHOJ-UHFFFAOYSA-N
Synonym sodium ethoxide, sodium ethylate, sodium ethanolate, sodiumethoxide, ethoxysodium, ethanol, sodium salt, caustic alcohol, naoet, etona, ethanol, sodium salt 1:1
PubChem CID 2723922
ChEBI CHEBI:52096
IUPAC Name sodium;ethanolate
SMILES CC[O-].[Na+]

Specifications

Density 0.868
Flash Point 20°C (68°F)
Refractive Index 1.3875
Linear Formula NaOCH2CH3
Quantity 100 mL
UN Number UN3274
Beilstein 3593646
Sensitivity Air and moisture sensitive
Merck Index 14,8613
Solubility Information Miscible with alcohol.
Formula Weight 68.05
Concentration or Composition (by Analyte or Components) 21% w/w in ethanol
Chemical Name or Material Sodium ethoxide
Show More Show Less
Hazard Category H226-H314-H335
Hazard Statement GHS H Statement
H225-H314
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Precautionary Statement P201-P202-P210-P233-P235-P235+P410-P240-P241-P242-P243-P260-P264b-P271-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ALCOHOLATES SOLUTION, N.O.S.
EINECSNumber 205-487-5
TSCA Yes
Recommended Storage Ambient temperatures

For Manufacturing and Laboratory Use. Not for beverage use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.