Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium ethoxide, 96%, Thermo Scientific™

Catalog No. AC168590010
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC168590010 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC168590010 Supplier Thermo Scientific Chemicals Supplier No. 168590010

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-52-6
Molecular Formula C2H5NaO
Molecular Weight (g/mol) 68.051
InChI Key QDRKDTQENPPHOJ-UHFFFAOYSA-N
Synonym sodium ethoxide, sodium ethylate, sodium ethanolate, sodiumethoxide, ethoxysodium, ethanol, sodium salt, caustic alcohol, naoet, etona, ethanol, sodium salt 1:1
PubChem CID 2723922
ChEBI CHEBI:52096
IUPAC Name sodium;ethanolate
SMILES CC[O-].[Na+]

Specifications

Melting Point 260.0°C
Linear Formula C2H5ONa
Quantity 1 kg
Solubility Information Solubility in water: reacts. Other solubilities: soluble in absolute alcohol without decomposition
Formula Weight 68.04
Grade Pure
Chemical Name or Material Sodium ethoxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.