Learn More
Electron Microscopy Sciences Sodium Ethylate 25 GR
SDP
Catalog No. 5028738
Encompass_Preferred
CAS #141-52-6 C₂H₅ONa F.W. 68.05
Sodium Ethoxide is used as an agent for flattening thick sections (0.5-2.0 micron) of epoxy embedded material. Felix Sierra, Christa Hegele-Hartung and Henning M. Beier (1990). Also see: An improved technique for flattening semithin sections of epoxy-embedded tissues used for high resolution light microscopy. J. of Microscopy Vol 159, Pt 1, (July 1990), pp. 95-98
Catalog No. 50-287-38 Supplier Electron Microscopy Sciences Supplier No. 21150
CAS #141-52-6 C₂H₅ONa F.W. 68.05
Sodium Ethoxide is used as an agent for flattening thick sections (0.5-2.0 micron) of epoxy embedded material. Felix Sierra, Christa Hegele-Hartung and Henning M. Beier (1990). Also see: An improved technique for flattening semithin sections of epoxy-embedded tissues used for high resolution light microscopy. J. of Microscopy Vol 159, Pt 1, (July 1990), pp. 95-98