Learn More
Medchemexpress LLC Sodium ethylparaben 100g
SDP
Catalog No. 5000205293
Encompass_Preferred
Sodium ethylparaben (Sodium ethyl 4-hydroxybenzoate) acts as a water soluble antiseptic usually employed as safe for cosmetics[1]
Catalog No. 50-002-05293 Supplier Medchemexpress LLC Supplier No. HYW099118100G
Sodium ethylparaben (Sodium ethyl 4-hydroxybenzoate) acts as a water soluble antiseptic usually employed as safe for cosmetics[1]