Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Ethylxanthate 97.0+%, TCI America™
SDP

Catalog No. E0195500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0195500G 500 g
E019525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E0195500G Supplier TCI America Supplier No. E0195500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 140-90-9
Molecular Formula C3H5NaOS2
Molecular Weight (g/mol) 144.182
MDL Number MFCD00068314
InChI Key RZFBEFUNINJXRQ-UHFFFAOYSA-M
Synonym Ethylxanthic Acid Sodium Salt, Sodium O-Ethyl Dithiocarbonate, Sodium Xanthogenate
PubChem CID 23690437
IUPAC Name sodium;ethoxymethanedithioate
SMILES CCOC(=S)[S-].[Na+]

Specifications

Color Yellow
Quantity 500 g
Formula Weight 144.18
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Ethylxanthate
EINECSNumber (2)-1287
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.