Learn More
Selleck Chemical LLC Sodium Etidronate S5198-25mg
SDP
Catalog No. 503108134
Encompass_Preferred
Sodium Etidronate (Didronel EHDP Etidronic acid) is a non-aminobisphosphonate that exhibits protein tyrosine phosphatase (PTP) inhibitory activity
Catalog No. 50-310-8134 Supplier Selleck Chemical LLC Supplier No. S519825MG
Sodium Etidronate (Didronel EHDP Etidronic acid) is a non-aminobisphosphonate that exhibits protein tyrosine phosphatase (PTP) inhibitory activity