Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium ferrocyanide decahydrate

Catalog No. AC459290050
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC459290050 5 kg
AC459292500 250 g
AC459290010 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC459290050 Supplier Thermo Scientific Chemicals Supplier No. 459290050
Add to Cart

CAS: 14434-22-1 | C6H20FeN6Na4O10 | 484.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14434-22-1
Molecular Formula C6H20FeN6Na4O10
Molecular Weight (g/mol) 484.06
MDL Number MFCD00149274
InChI Key JFSUDVTVQZUDOP-UHFFFAOYSA-N
SMILES O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[Na+].[Na+].[Fe++].[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N

Specifications

Quantity 5 kg
Solubility Information Solubility in water: 369g/L (20°C). Other solubilities: insoluble in alcohol
Formula Weight 484.06
Loss on Drying 1% max. (free moisture)
Chemical Name or Material Sodium ferrocyanide decahydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.