Learn More
Apexbio Technology LLC sodium fluoride 10000mg
SDP
Catalog No. 5000087672
Encompass_Preferred
Sodium fluoride is an inorganic compound widely utilized in biomedical research for its inhibitory action on enzymes involved in cellular metabolism particularly enolase within the glycolytic pathway By inhibiting glycolytic activity sodium fluoride disrupts ATP production making it a valuable tool for assessing metabolic processes in vitro It is commonly employed to prevent glycolysis in blood samples for biochemical assays and in studies investigating metabolic regulation cellular energetics and mechanisms of fluoride toxicity Sodium fluoride s modulation of metabolic enzymes supports its use in mechanistic studies of cell physiology and metabolic control
Catalog No. 50-000-87672 Supplier Apexbio Technology LLC Supplier No. B784610000
Sodium fluoride is an inorganic compound widely utilized in biomedical research for its inhibitory action on enzymes involved in cellular metabolism particularly enolase within the glycolytic pathway By inhibiting glycolytic activity sodium fluoride disrupts ATP production making it a valuable tool for assessing metabolic processes in vitro It is commonly employed to prevent glycolysis in blood samples for biochemical assays and in studies investigating metabolic regulation cellular energetics and mechanisms of fluoride toxicity Sodium fluoride s modulation of metabolic enzymes supports its use in mechanistic studies of cell physiology and metabolic control